Learn More
Cytiva ExoProStar™ S PCR and Sequence Reaction Clean-up Kit
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
Marca: Cytiva US79050
Dettagli aggiuntivi : CAS : 56-81-5 Peso : 0.10000kg
Descrizione
- Easy to automate
- Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
- Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
- Scalable for different reaction sizes
- No loss of PCR product
- Complete heat inactivation of the enzymes within 10 minutes
- Fast 15 minute protocol
- Updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
- Improves digestion efficiency with no degradation of the target PCR product
- Sample is completely intact and ready for immediate use in manual or automated processes
Specifica
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes. | |
PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product | |
500 | |
illustra ExoProStar S |