Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity
For one free-selectable temperature and humidity value, and measuring of light intensity
Marca: Memmert™ HPP110ecoD00137
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Includes temperature distribution survey at Memmert
- For 27 measuring points to DIN 12880:2007-05
- PQ check list as support for validation by customer.
Specifica
HPP110eco | |
IQ/OQ Document with Device-specific Works Test Data |