Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum is a gas collecting tube with O-ring valves on each end and a centrally located septum.
Marca:  GPE Scientific™ GSS/500
Dettagli aggiuntivi : Peso : 0.30000kg
Codice prodotto. 15824774
  • 192.00€ / Pezzo
Spedizione stimata: 30-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Cylindrical, oblong shape
  • Gas tight container
  • Gauged volume
  • Standard collecting tube with valved ends
  • Includes centralized septum
Specifica
Specifica
Gas Sample Tube
500 mL