Learn More
Justrite™ Drum Faucets, Extender, and Valve
Self-closing safety drum faucets
Marca: Justrite™ 08910
Dettagli aggiuntivi : Peso : 0.52000kg
Descrizione
- Self-closing brass safety faucets feature a PTFE fluorocarbon resin seal, flame arrester and standard 0.75 in. NPT thread
- Brass faucets screw into drum; adjustable model adjusts to spout-down position
- Flexible faucet extensions provide accuracy, prevent spills; available in 8 in. long (20.3cm) brass and 6 in. long (15.2cm) stainless steel
- Brass drum gate valve fits 2 in. (5.1cm) NPT openings and has wide mouth for easy dispensing of viscous liquids from drums in horizontal position
Certifications
Faucets and valve are FM approved.
Specifica
Adjustable Drum Faucet | |
FM | |
Brass | |
PTFE Fluorocarbon Resin seal, internal flame arrester and |