missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10514-2-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896943

  • 121.38€ / 20µL

Spedizione stimata: 04-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
Specifica

Specifica

Kallikrein 5
Polyclonal
Unconjugated
KLK5
Kallikrein 5, Kallikrein like protein 2, kallikrein related peptidase 5, KLK L2, KLK5, KLKL2, SCTE, Stratum corneum tryptic enzyme
Rabbit
Antigen Affinity Chromatography
RUO
25818
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y337
KLK5
Kallikrein 5 Fusion Protein Ag0799
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali