Learn More
KAT2B Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 28770-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00020kg
Codice prodotto. 17221416
  • 399.00€ / 150µL
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
The KAT2B antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human KAT2B. This antibody recognizes human antigen. The KAT2B antibody has been validated for the following applications: WB,ELISA analysis.
Specifica
Specifica
KAT2B
Polyclonal
Unconjugated
150 μL
8850
Liquid
Western Blot, ELISA
700 μg/mL