Learn More
KAT2B Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28770-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
The KAT2B antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human KAT2B. This antibody recognizes human antigen. The KAT2B antibody has been validated for the following applications: WB,ELISA analysis.
Specifica
KAT2B | |
Polyclonal | |
Unconjugated | |
20 μL | |
8850 | |
Liquid |
Western Blot, ELISA | |
700 μg/mL | |
Rabbit | |
Primary | |
Human |