missing translation for 'onlineSavingsMsg'
Learn More

KCNQ2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1D4A1, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66774-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898955

  • 399.00€ / 150µL
Spedizione stimata: 21-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system and KNCQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.
Specifica

Specifica

KCNQ2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43526, O88943, Q9Z351
Kcnq2
KCNQ2 Fusion Protein Ag25781
150 μL
Primary
Human, Mouse, Porcine, Rat
Antibody
IgG1
Western Blot
1D4A1
Unconjugated
KCNQ2
BFNC, EBN, EBN1, ENB1, HNSPC, KCNA11, KCNQ2, KQT like 2, KV7.2, KVEBN1
Mouse
Protein G
RUO
110257402, 16536, 170848, 3785
-20°C
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali