Learn More
VACUUBRAND™ KF DN 10mm Stainless Steel Small Flange With Female Ground Joint
Marca: VACUUBRAND™ 20662801
Dettagli aggiuntivi : Peso : 0.50000kg
Specifica
Stainless Steel | |
NS 19/38 | |
Small Flange |
1 Each | |
KF DN 10 mm |