Learn More
DWK Life Sciences Kimble™ Glass Racks for Westergren Tubes
Reduces the chance of bio hazardous contamination
Marca: DWK Life Sciences 2215N
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Racks feature leveling bubbles and adjustment screws
- Tubes are held firmly in a perpendicular position to ensure accurate readings.
Specifica
Not Autoclavable | |
Glass | |
10 | |
For westergren and wintrobe tubes |