Learn More
KMT2C Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 28437-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00020kg
Codice prodotto. 17251406
  • 140.00€ / 20µL
Spedizione stimata: 20-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
The KMT2C antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human KMT2C. This antibody recognizes human antigen. The KMT2C antibody has been validated for the following applications: WB,ELISA analysis.
Specifica
Specifica
KMT2C
Polyclonal
Unconjugated
20 μL
58508
Liquid
Western Blot, ELISA
500 μg/mL