Learn More
Epredia™ Lab Vision™ Rhodopsin (Opsin) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Provide accurate, reproducible results with the Epredia™ Rhodopsin (Opsin) Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-1233-P
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
Rhodopsin is a glycoprotein which is a visual pigment in mammals, birds, amphibians, and reptiles. The rhodopsin protein is present on the photoreceptor cells in the retina.Host Species: Mouse
Clone: RET-P1
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Membrane preparation from adult rat retina
Molecular Weight: 39kDa
Positive Control: Brain or retina
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifica
Rhodopsin (Opsin) Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Cancer and Tumor Biology | |
Human, Mouse, Rat | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
RET-P1 | |
Unconjugated | |
Membrane preparation from adult rat retina | |
RUO | |
Primary | |
Concentrated |