missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGALS3BP Mouse anti-Human, Clone: 2D8E11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60066-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.Specifica
LGALS3BP | |
Monoclonal | |
1.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q08380 | |
LGALS3BP | |
LGALS3BP Fusion Protein Ag0294 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
2D8E11 | |
Unconjugated | |
LGALS3BP | |
90K, BTBD17B, Galectin 3 binding protein, LGALS3BP, M2BP, Mac 2 binding protein, MAC 2 BP, MAC2BP, Tumor associated antigen 90K | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
3959 | |
-20°C | |
Liquid |