Learn More
Lenz Laborglasintrumente Long Ground Cones With Tip
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 01100819
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Conical ground cones
- Made of DURAN™ tubing
Specifica
10/Pk. | |
1.8 mm | |
120 mm |
NS 19/38 | |
17 mm | |
Borosilicate Glass (3.3) |