Learn More
Lovibond™ Aqua Comparitor Color Discs: pH
For testing water with the Aqua Comparator
Marca: Lovibond™ 146130
Dettagli aggiuntivi : Peso : 0.02900kg
Descrizione
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for pH using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifica
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
4 to 10 pH | |
Universal pH Color Disc |