Learn More
Lovibond™ Borosilicate Glass Colorimetry Cells
Precision fused cells
Marca:  Lovibond™ 656010
Dettagli aggiuntivi : Peso : 0.02000kg
Codice prodotto. 15869791
  • 139.00€ / Pezzo
Spedizione stimata: 28-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Available in a range of dimensions and path lengths, made from optical glass to the highest standards. Note: For instruments equipped with a heater unit and whenever cells are subjected to thermal shock, it is recommended that borosilicate cells be used.
Series: W600 Borosilicate
Specifica
Specifica
Colorimetry Cell
Model Fs, PFX195, 880, 950 and 995 instruments with heated Samples (up to 100°C)
Borosilicate Glass
33 mm