Learn More
Cole-Parmer™ Sacchetti in polietilene a bassa densità
Store a variety of solids or powders in these LDPE bags. Low Density Polyethylene Bags stand up to your storage needs with a strong bottom seal.
Marca: Cole-Parmer™ 06505-12
Dettagli aggiuntivi : Peso : 5.21000kg
Descrizione
- Seal with heat sealer or bag ties
- Good for general laboratory storage
- Available in a variety of sizes and thicknesses
Specifica
Low-Density Bag | |
Polyethylene | |
3 mil | |
457 mm |
Clear | |
500 Each | |
457 x 152mm | |
152 mm |