Learn More
Sartorius Stedim Biotech™ Malt Extract NPS Including Dispenserfilter
Sterile, dehydrated culture media pads with a gridded membrane filter.
Marca:  Sartorius Stedim Biotech™ 14050--50----RDN
240.00 EUR valido fino al 2024-12-31
Usa il codice promo "23270" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.31000kg
Codice prodotto. 15912490
  • 262.00€ / pezzi 100
Spedizione stimata: 07-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Malt extract nutrient pads with gridded membranes
  • For determination of yeasts and molds acc. to international Standards e.g. APHA, AOAC, IFU and internal SOPs.
  • Trouble-free storage up to 24 months
  • Economical by time- and cost-savings
  • Highly versatile -multiple applications
  • Consistent quality
  • Validation guide available
  • Easy handling
  • Compliance with international standards and references
Specifica
Specifica
Gray
Malt Extract
Yeast, Molds
10 x 10 petri dishes with NPS in bags, dispenser-ready membrane filters
100/Pk.
0.45μm Pore Size Disc
Sterile
White
0.45μm
50 mm