Learn More
Emma Safety Footwear Mammoet M1966 D Safety Shoes
State-of-the art Sympatex™ construction provides perfect foot protection for 'hands on' roles within the Chemical Industry and Agriculture. Virtually waterproof, these boots allow feet to breathe properly, keeping them cool and dry.
Marca: Emma Safety Footwear 584848-45
Dettagli aggiuntivi : Peso : 2.41400kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Duo Heat Resistant Outsole
- Sympatex™ Waterproof
- Heat and cold insulated
- Sanitized
Specifica
EN ISO 20345:2011 | |
Mammoet M1966 D | |
Oiled Nubuck | |
Steel |
Black | |
Hydro-Tec™ Sanitized Silver / Sympatex™ | |
45 |