Learn More
Emma Safety Footwear Mammoet M2001 XD Safety Shoes
A perfectly integrated steel toe protects the feet against falling pieces of metal and other exterior threats. Hydro-Tec™ Sanitized Silver removes moisture and provides optimum ventilation. Ankle protection Comfort Sponge with added padding provides a comfortable and safe feel.
Marca: Emma Safety Footwear 745868-42
Dettagli aggiuntivi : Peso : 1.99500kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Heat and cold insulated
- Sanitized
Specifica
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
42 |
Laces | |
Mammoet M2001 XD | |
Oiled Nubuck | |
Steel |