Learn More
Abeba™ Manager 3110 Shoes
All calf leather construction. ESD dissipative.
Marca: Abeba™ 3110/43
Dettagli aggiuntivi : Peso : 1.20000kg
Descrizione
- CE
- Calf leather
- Black
- Antistatic leather outsole sole
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
- Leather insole
Specifica
Black | |
Leather | |
Leather |
Microfiber | |
43 | |
3110 |