Learn More
Emma Safety Footwear Melvin MF Safety Shoes
Keep your shoes on whenever you enter the factory or go through security gates. The metal-free shoe saves you plenty of time over the course of the day. The microfiber top is easily maintained, supports the foot properly and is amazingly soft, supple and comfortable.
Marca: Emma Safety Footwear MM43864746
Dettagli aggiuntivi : Peso : 1.78600kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Microfber construction
- TPU GripForce™ Easy Twist Outsole
- Metal-free
- Chrome-free lining
- 3M™ Reflective Striping
- ESD certified
- Hydro-Tec™ Sanitized Silver
Specifica
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Antibacterial | |
46 |
Laces | |
Melvin MF | |
Microfiber | |
Composite |