Learn More
Metro™ Double-Deep qwikTRAK™ Stationary End Unit Kits
Connects end units to tracks and anchors tracks to floor
Marca: Metro™ LBTEK3
Dettagli aggiuntivi : Peso : 26.85600kg
Descrizione
- For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ and MetroMax iQ shelving systems
- Kit includes 74 in. (188cm) high stationary posts and connecting hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifica
74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary end units to track and anchors track to floor | |
Stationary End Unit Kit | |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system | |
Steel |
33.5 kg | |
73.6 lb. | |
Metroseal Green Epoxy |