Learn More
Metro™ MetroMax™ iQ Polymer Shelving Accessory, Shelf Ledge, Stackable
2 in. ledges provide a secure lip to secure items during transport or to meet seismic or other safety requirements
Marca: Metro™ MXLS18-2S
Dettagli aggiuntivi : Peso : 0.20000kg
Descrizione
Easily attach with no tools to MetroMax i solid or open grid shelves. Other sizes and styles available.
- Snap on and off quickly
- Easily stacked to contain larger items
- Corrosion proof
- Ships assembled
Specifica
18 in. | |
2 in. | |
45.7 cm |
5 cm | |
Metro™ MetroMax i™ and Q™ shelving | |
Stainless Steel |