missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGMT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17195-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
O6-Methylguanine-DNA Methyltransferase (MGMT) is an important DNA repair protein involved in tumor cell resistance to the cytostatic activity of chemotherapeutic alkylating agents. This protein is also effective in protecting normal cells against the genotoxic and carcinogenic effects of DNA alkylation. The alkylating drug resistance is caused by MGMT′s ability to remove DNA alkyl groups introduced in the O6 position of guanine. MGMT is expressed in highly variable amounts, depending upon the cell and tissue type, species, and cellular growth characteristics. In addition, MGMT activity varies among groups of tumors and within a particular type of tumor.Specifica
MGMT | |
Polyclonal | |
Unconjugated | |
MGMT | |
MGMT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4255 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16455 | |
MGMT | |
MGMT Fusion Protein Ag9996 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |