Learn More
ROBU VitraPOR™ Micro-Filter Funnel, According to Pregl
Micro-Filter Funnel for small volumes.
Marca: ROBU VitraPOR™ 28041
Dettagli aggiuntivi : Peso : 0.06460kg
Descrizione
- Highly resistant to most chemicals and temperatures up to 500°C
Specifica
4 mL | |
Borosilicate Glass 3.3 | |
1 | |
5/Pk. |