Learn More
Cytiva (formerly Pall Lab) MicroFunnel™ Filters Funnel with Metricel™ Black Membrane
Ready-to-use, sterile and individually packaged MicroFunnel filter funnels reduce chances of cross contamination.
Marca: Cytiva (formerly Pall Lab) 4817
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
- Easy squeeze removal of funnel to access membrane – no twisting means less RSI and membrane tearing
- Proven membrane performance
- For research use only
Specifica
300 mL | |
0.45 μm | |
Sterile |
Black | |
20/Pk. | |
Metricel Membrane Modified PES |