Learn More
3M™ Mini Flip-Top Vial
Leak-resistant plastic cylinder with a flip-open lid that seals securely for safe shipping and handling.
Marca:  3M™ 7000053834
Dettagli aggiuntivi : CAS : 7732-18-5 Peso : 0.34500kg
Codice prodotto. 17263734
  • 183.00€ / pezzi 100
Spedizione stimata: 21-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Hinged lid can be opened with one hand and stays open for easy sample additions.
  • Decontaminated through gamma irradiation and ready for use with no additional blending required.
  • Flat bottom tube stands on its own, no holder required.
  • Leak resistant vial helps prevent leakage during shipping.
  • 2 year shelf life from date of manufacture provides long-term readiness.
Specifica
Specifica
9 mL Buffered Peptone Water Broth
100/Cs.