Learn More
Cytiva (formerly Pall Lab) Minimate™ Tangential Flow Filtration (TFF) Capsule with Omega™ Membrane
Concentration and diafiltration (desalting or buffer exchange) processes can be performed on the same system with minimal user intervention.
Marca: Cytiva (formerly Pall Lab) OA050C12
Dettagli aggiuntivi : Peso : 0.45400kg
Descrizione
- Efficient processing
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
PES | |
Non-sterile | |
1/Pk. |
50 kDa | |
50 cm2 |