missing translation for 'onlineSavingsMsg'
Learn More

MMP21 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 55289-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896775

  • 121.38€ / 20µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This antibody is specific to MMP21.

This gene encodes a member of the matrix metalloproteinase family. Proteins in this family are involved in the breakdown of extracellular matrix for both normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, and disease processes, such as asthma and metastasis. The encoded protein may play an important role in embryogenesis, particularly in neuronal cells, as well as in lymphocyte development and survival.
Specifica

Specifica

MMP21
Polyclonal
Unconjugated
MMP21
matrix metallopeptidase 21, Matrix metalloproteinase 21, MMP 21, MMP21
Rabbit
Antigen Affinity Chromatography
RUO
118856, 214766
-20°C
Liquid
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K3F2, Q8N119
Mmp21
Peptide
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali