missing translation for 'onlineSavingsMsg'
Learn More

MOPSO, 0.2M buffer soln., pH 7.0, Thermo Scientific™

Catalog Number 15414069
Click to view available options
Quantità:
100 mL
250 mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
79803-73-9
C7H14NNaO5S
247.241
MFCD00070007
WSFQLUVWDKCYSW-UHFFFAOYSA-M
23675131
sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate
C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+]
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
79803-73-9
C7H14NNaO5S
247.241
MFCD00070007
WSFQLUVWDKCYSW-UHFFFAOYSA-M
23675131
sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate
C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico MOPSO buffer soln.
Concentrazione 0.2 M
pH 7.0
Forma fisica Liquid
Quantità 100 mL
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'