Learn More
MP Biomedicals™ Alkaline Phosphatase, Powder
From calf intestine
Marca: MP Biomedicals™ 0219068001
66.35 EUR valido fino al 2024-12-31
Usa il codice promo "23257" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : CAS : 9001-78-9 Peso : 0.00100kg
Descrizione
Activity: ∽1.0U/mg. Unit Definition: One unit causes hydrolysis of one micromole of p-nitrophenyl phosphate per minute at pH 9.6 and 25°C. Freeze-dried powder.
Specifica
≥1 U/mg | |
9.8 | |
-20°C | |
1 kU | |
140000 | |
Lyophilized Powder |
Off-White | |
Used to remove 5'-phosphate groups from Nucleic Acid molecules to prevent self-ligation of DNA fragments and vectors during recombinant DNA manipulation | |
Calf intestine | |
Alkaline Phosphatase | |
Acidification to 4.5 pH reversibly inactives the Enzyme, Inorganic Phosphate, chelating agents, EDTA |