missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFR Mouse anti-Human, Clone: 1F7A3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66612-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.Specifica
MTHFR | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P42898 | |
MTHFR | |
MTHFR Fusion Protein Ag14970 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
1F7A3 | |
Unconjugated | |
MTHFR | |
MTHFR | |
Mouse | |
Protein A | |
RUO | |
4524 | |
-20°C | |
Liquid |