missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTRF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11581-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain non-cognate termination stop codons AGG and AGA.Specifica
MTRF1 | |
Polyclonal | |
Unconjugated | |
MTRF1 | |
MRF 1, MRF1, MtRF 1, MTRF1, MTTRF1, RF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
211253, 9617 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75570, Q8K126 | |
MTRF1 | |
MTRF1 Fusion Protein Ag2170 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |