missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH2 Mouse anti-Human, Clone: 1F1B6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66212-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The antibody is specific to MYH2 and will not bind other myosins.
Muscle contraction. Required for cytoskeleton organization.Specifica
MYH2 | |
Monoclonal | |
0.9 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UKX2 | |
MYH2 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
1F1B6 | |
Unconjugated | |
MYH2 | |
MYH2, MYH2A, MYHas8, MyHC 2A, MyHC IIa, MYHSA2, Myosin 2, Myosin 2a, Myosin heavy chain 2, Myosin heavy chain 2a, Myosin heavy chain IIa | |
Mouse | |
Protein A | |
RUO | |
4620 | |
-20°C | |
Liquid |