Learn More
MyoD1 Ab-2 Mouse Monoclonal Antibody, Epredia™
Deliver accurate, reproducible results with the Epredia™ MyoD1 Ab-2, Mouse Monoclonal Antibody.
Marca: Epredia MS-278-PABX
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
Research Areas: Muscle Markers; MyoD1 is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occassionally nuclear expression of MyoD1 is seen in ectomesenchymoma and a subset of Wilm's tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing's sarcomas and alveolar soft part sarcomas.Host Species: Mouse
Clone: 5.2F
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Chicken. Others not known.
Epitope: aa 3-56
Immunogen: Recombinant mouse MyoD1 protein
Molecular Weight: 45kDa
Positive Control: SW80 cells
Cellular Localization: Nuclear
Recommended for:
- Immunofluorescence
- Immunoprecipitation (Native verified)
- Electron Microscopy
Note:
USA: RUO; Int'l: RUO
Specifica
MyoD1 Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Muscle Biology | |
Chicken, Human, Mouse, Rat | |
IgG2a κ |
Immunofluorescence, Immunoprecipitation | |
5.2F | |
Unconjugated | |
Recombinant mouse MyoD1 protein | |
RUO | |
Primary | |
Concentrated |