missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55462-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifica
Myosin Light Chain 2 | |
Polyclonal | |
Unconjugated | |
MYL2 | |
CMH10, MLC 2, MLC 2v, MLC2, MLC2v, MYL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17906, 363925, 4633 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.79 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08733, P10916, P51667 | |
Myl2 | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |