Learn More
Hamilton™ Non-Filtered Low Retention Racked Pipette Tips
With surface walls completely free of occlusions and cavities so fluid retention is virtually eliminated.
Marca: Hamilton™ 68008-21
Dettagli aggiuntivi : Peso : 0.20000kg
Descrizione
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Available pre-sterilized or non-sterile
- 96 tips to a rack
- Delivering outstanding accuracy and precision, guaranteed
Specifica
200 μL | |
Non-sterile | |
Virgin Polypropylene |
Non-Filtered | |
10 racks; 960 total tips |