missing translation for 'onlineSavingsMsg'
Learn More

NPM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11960-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899373

  • 121.38€ / 20µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The protein encoded by this gene is related to the nuclear chaperone phosphoproteins, nucleoplasmin and nucleophosmin. This protein is strongly expressed in diverse cell types where it localizes primarily to the nucleus. Based on its similarity to nucleoplasmin and nucleophosmin, this protein likely functions as a molecular chaperone in the cell nucleus.
Specifica

Specifica

NPM3
Polyclonal
Unconjugated
NPM3
NPM3, nucleophosmin/nucleoplasmin, 3, Nucleoplasmin 3, PORMIN, TMEM123
Rabbit
Antigen Affinity Chromatography
RUO
10360, 18150, 502389
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75607, Q9CPP0
Npm3
NPM3 Fusion Protein Ag2564
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali