missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRBP2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4F10H7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66687-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Nrbp2 may regulate apoptosis of neural progenitor cells during their differentiation.Specifica
NRBP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q91V36, Q9NSY0 | |
NRBP2 | |
NRBP2 Fusion Protein Ag17011 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
4F10H7 | |
Unconjugated | |
NRBP2 | |
DKFZp434P086, NRBP2, pp9320, TRG 16, TRG16 | |
Mouse | |
Protein A | |
RUO | |
110260412, 223649, 340371, 680451 | |
-20°C | |
Liquid |