missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTF2 Mouse anti-Human, Clone: 4H7F9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66063-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The MW of this protein is 14 kDa, and this antibody specifically recognizes the 14 kDa protein.
The protein encoded by this gene is a cytosolic factor that facilitates protein transport into the nucleus. It interacts with the nuclear pore complex glycoprotein p62. This encoded protein acts at a relative late stage of nuclear protein import, subsequent to the initial docking of nuclear import ligand at the nuclear envelope. It is thought to be part of a multicomponent system of cytosolic factors that assemble at the pore complex during nuclear import.Specifica
NTF2 | |
Monoclonal | |
0.86 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61970 | |
NUTF2 | |
NTF2 Fusion Protein Ag7879 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
4H7F9 | |
Unconjugated | |
NUTF2 | |
NTF 2, NTF2, nuclear transport factor 2, NUTF 2, NUTF2, Placental protein 15, PP15 | |
Mouse | |
Protein G | |
RUO | |
10204 | |
-20°C | |
Liquid |