missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleocapsid protein Mouse anti-SARS-CoV-2, Unconjugated, Clone: AS46, AcroBiosystems
Mouse Monoclonal Antibody
Marca: ACROBiosystems NUN-S46A1-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Nucleocapsid protein Monoclonal antibody specifically detects Nucleocapsid protein in SARS-CoV-2 samples.Specifica
Nucleocapsid protein | |
AS46 | |
Human (HEK293) | |
RUO | |
SARS-CoV-2 | |
IgG1 |
Monoclonal | |
Unconjugated | |
100 μg | |
Primary | |
-20°C |