Learn More
Masterflex™ Nylon Barbed Tee Connector
Designed for quick and easy assembly without tools
Marca: Masterflex™ 30623-63
Dettagli aggiuntivi : Peso : 0.22000kg
Descripción
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Especificaciones
Barbed Tee Connector | |
Not Autoclavable | |
4 in. | |
1 in. | |
1 in. | |
2 1/2 in. | |
Translucent |
Barb Connections | |
Nylon | |
1 in. | |
1 in. | |
Ethylene Oxide | |
T-shape |