Learn More
Brady™ Nylon Cloth Printable Labels
Blank, printable nylon cloth labels for Laser Printers.
Marca: Brady™ 029684
Dettagli aggiuntivi : Peso : 0.09750kg
Descrizione
- Blank, printable nylon cloth labels for Laser Printers.
Specifica
Halogen Free (DIN VDE 0472 Part 815) | |
Permanent Acrylic, Permanent | |
Matte | |
Laser | |
General Identification, Laboratory labelling, Wire and cable labelling | |
15.24 mm | |
-40°C | |
1092 Pk. | |
12.7 x 19.05 mm (W x H) | |
-40°C to +90°C | |
0.0065 in. | |
19.05 x 12.7 mm | |
19.05 mm | |
12.7 mm |
LaserTab Laser Printable Labels | |
White | |
Nylon | |
Labels | |
Laser Printers | |
13 | |
1092 Labels/Pk. | |
Rectangle | |
-40°F to +194°F | |
80 lb./in. | |
0.165 mm | |
0.75 in. | |
0.5 in. |