Learn More
DWK Life Sciences O-Ring, transparent, from silicone (VMQ), not suitable for desiccators
Seal is suitable for the tight sealing of flat flange vessels with annular groove.
Marca:  DWK Life Sciences 292255101
114.00 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.03000kg
Codice prodotto. 13497068
  • 120.00€ / pezzi 5
Spedizione stimata: 06-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Flat flange vessels are large-volume reaction vessels consisting of a lower part and a lid. These two parts are held together by a clamping ring. The transparent O-ring made of silicone serves as a seal for flat flange vessels with an annular groove. It prevents tilting of the ground surfaces between the lower part and the lid and convinces by its good elasticity. The chemical resistance is limited compared to FEP coated O-rings.
  • Accessories for flat flange vessels from silicone (VMQ), so is highly elastic
  • The chemical resistance of silicone is lower than FEP coated O-rings
Specifica
Specifica
Tight closure of flange vessels
5/Pk.
Silicone (VMQ)
133 mm