Learn More
Carl Zeiss™ Objective A Achromat
Use these universal objectives for routine research. Zeiss™ A-Plan Objectives provide rich contrast and can be used with eyepieces with a field of view up to 23mm.
Marca: Carl Zeiss™ 421020-9900-000
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- This class of objectives provides an attractive alternative to Achroplan objectives, both in routine and research applications
- Allow contrasting methods ranging from brightfield to phase contrast and for excitation wavelength within the visible spectrum
- Can be used for epifluorescence work
Specifica
Obiettivo A | |
Numerical Aperture: 0.06 mm | |
2.5X | |
1 Ea. |