Learn More
Carl Zeiss™ Objective A Achromat
Use these universal objectives for routine research. Zeiss™ A-Plan Objectives provide rich contrast and can be used with eyepieces with a field of view up to 23mm.
Marca:  Carl Zeiss™ 421060-9900-000
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11855083
  • 588.10€ / Pezzo
Spedizione stimata: 25-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • This class of objectives provides an attractive alternative to Achroplan objectives, both in routine and research applications
  • Allow contrasting methods ranging from brightfield to phase contrast and for excitation wavelength within the visible spectrum
  • Can be used for epifluorescence work
Specifica
Specifica
Obiettivo A
Numerical Aperture: 0.65 mm
40X
1 Ea.