missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10867-2-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Description
Oct2 is a transcription factor that specifically binds to the octamer motif (5′-ATTTGCAT-3′). Oct2 regulates transcription in a number of tissues in addition to activating immunoglobulin gene expression. It also modulates transcription transactivation by NR3C1, AR and PGR. Oct2 is B-cell specific, belongs to the POU transcription factor family class-2 subfamily and contains 1 homeobox domain.Specifications
OCT2 | |
Polyclonal | |
Unconjugated | |
POU2F2 | |
Oct 2, OCT2, Octamer binding protein 2, OTF 2, OTF2, POU class 2 homeobox 2, POU2F2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18987, 5452 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09086, Q00196 | |
Pou2f2 | |
OCT2 Fusion Protein Ag0955 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |