missing translation for 'onlineSavingsMsg'
Learn More

OCT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10867-2-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897533

  • 325.38€ / 150µL

Spedizione stimata: 10-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Oct2 is a transcription factor that specifically binds to the octamer motif (5′-ATTTGCAT-3′). Oct2 regulates transcription in a number of tissues in addition to activating immunoglobulin gene expression. It also modulates transcription transactivation by NR3C1, AR and PGR. Oct2 is B-cell specific, belongs to the POU transcription factor family class-2 subfamily and contains 1 homeobox domain.
Specifica

Specifica

OCT2
Polyclonal
Unconjugated
POU2F2
Oct 2, OCT2, Octamer binding protein 2, OTF 2, OTF2, POU class 2 homeobox 2, POU2F2
Rabbit
Antigen Affinity Chromatography
RUO
18987, 5452
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P09086, Q00196
Pou2f2
OCT2 Fusion Protein Ag0955
150 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali