missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oligophrenin 1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11076-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Stimulates GTP hydrolysis of members of the Rho family. Its action on RHOA activity and signaling is implicated in growth and stabilization of dendritic spines, and therefore in synaptic function. Critical for the stabilization of AMPA receptors at postsynaptic sites. Critical for the regulation of synaptic vesicle endocytosis at presynaptic terminals. Required for the localization of NR1D1 to dendrites, can suppress its repressor activity and protect it from proteasomal degradation.Specifica
Oligophrenin 1 | |
Polyclonal | |
Unconjugated | |
OPHN1 | |
MRX60, oligophrenin 1, OPHN1, OPN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4983, 94190 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60890, Q99J31 | |
OPHN1 | |
Oligophrenin 1 Fusion Protein Ag1536 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |